Would you like to log in again?
Tissue-Tek® FormaGO® Formalin Neutralizer, Sakura® Finetek
Supplier: Sakura
9151
10852-346CS 256.8 USD InStock
10852-346
Tissue-Tek® FormaGO® Formalin Neutralizer, Sakura® Finetek
Spill Control Materials
Each pre-measured pouch of Tissue-Tek® FormaGO® Formalin Neutralizer neutralizes one gallon of 10% formalin in approximately 5 minutes
There are no drain-clogging solids after neutralizing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty