Would you like to log in again?
Standard Ferrules for PerkinElmer GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: RESTEK CORP
20212 27065 20231 27064
10852-528PK 244.09 USD InStock
10852-528 10852-526 RK20231 RK20212
Standard Ferrules for PerkinElmer GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Standard ferrules fit ID MXT® columns with internal diameters of 0.25 mm or 0.32 mm
  • VG2, 60% Vespel®/40% graphite blend offers the best combination of sealing and ease of workability
  • VG1, 85% Vespel®/15% graphite blend offers equivalent composition to Agilent Vespel®/graphite ferrules
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Stable to 400°C
  • Recommended for mass spec transfer lines
For use with PerkinElmer GCs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty