Would you like to log in again?
Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: RESTEK CORP
20205 27080 20255
RK20205PK 48.99 USD InStock
RK20205 10852-532 RK20255
Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Fits columns with internal diameters of 0.25 mm or 0.32 mm
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
For use with SRI and Buck Scientific GCs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty