Would you like to log in again?
Compact Ferrules for Agilent GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20249 27073 21008 21007 20248 27072
RK20249PK 399.69 USD InStock
RK20249 RK21008 10852-546 10852-548 RK21007 RK20248
Compact Ferrules for Agilent GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Compact ferrules are ideal for use with Agilent GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite or Vespel®/graphite material
  • Stable to 400°C
Ferrules feature internal diameters of 0.5 mm, compatible with columns 0.32 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty