Would you like to log in again?
Compact Ferrules for Agilent GCs, Fit 0.53 mm int.Ø MXT® Columns, Restek
Supplier: Restek
20252 20253 20263 27074 20264 27075
10803-418PK 68.61 USD InStock
10803-418 RK20253 RK20264 RK20252 RK20263 10852-550
Compact Ferrules for Agilent GCs, Fit 0.53 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Compact ferrules are ideal for use with Agilent GCs.
  • Durable, leak-tight
  • Preconditioned to eliminate out-gassing
  • Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
  • Recommended for mass spec transfer lines
Ferrules feature internal diameters of 0.8 mm, compatible with columns 0.45 or 0.53 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty