Would you like to log in again?
Ferrules for Thermo Scientific GCs, Fit 0.45/0.53 mm int.Ø MXT® Columns, Restek
Supplier: RESTEK CORP
20284 20285
10852-554PK 31.45 USD InStock
10852-554 10057-904
Ferrules for Thermo Scientific GCs, Fit 0.45/0.53 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
For use with Thermo Scientific GCs
  • Preconditioned to eliminate outgassing
  • High-purity, high-density graphite
  • Stable to 450°C
These ferrules contain no binders that can off-gas or adsorb analytes. Suitable for columns with internal diameters of 0.45 - 0.53 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty