Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Standard Ferrules, Fit 0.28 mm int.Ø MXT® Columns, Restek

Supplier: RESTEK CORP
20232 27066
10852-536PK 63.15 USD InStock
10852-536 10852-564
Standard Ferrules, Fit 0.28 mm int.Ø MXT® Columns, Restek
Fittings and Connectors
Standard ferrules fit 0.28 mm ID MXT® columns.

  • Seal with minimal torque
  • Reusable
  • Preferred for vacuum and high-pressure areas
  • Stable to 400°C

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty