Would you like to log in again?
Ferrules for Micropacked Columns, Capillary—for ¹/₁₆" Compression-Type Fittings, Restek
Supplier: RESTEK CORP
27069 22218 21060 24912 21058 22217
10852-612PK 48 USD InStock
10852-612 10852-598 10852-542 10852-566 10852-610 10852-594
Ferrules for Micropacked Columns, Capillary—for ¹/₁₆" Compression-Type Fittings, Restek
Fittings and Connectors
Ferrules are designed for use with micropacked columns, with a capillary design ideal for 1/16" compression-type fittings
  • Graphite construction
  • 1/16" fitting size
Fit columns with internal diameters of 0.75 mm.
Ordering information: For micropacked columns.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty