Would you like to log in again?
Capillary Column Caps, Restek
Supplier: Restek
22858 21044
10852-844EA 137.96 USD InStock
10852-844 10852-878
Capillary Column Caps, Restek
Fittings and Connectors
Silicone or glass capillary column caps can be attached to the column in seconds to form an airtight seal.
  • Increase column lifetime—prevent moisture and air from entering the column during storage
  • Two styles to choose from: glass or silicone material
  • Glass caps are color-coded for identifying detector and injector ends
  • Not recommended for reuse
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty