Would you like to log in again?
Spanner Wrench for Agilent 5890/6850/6890/7890 FID Collector Assemblies, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22329
10852-848EA 94.78 USD InStock
10852-848
Spanner Wrench for Agilent 5890/6850/6890/7890 FID Collector Assemblies, Restek
Wrenches
Use a spanner wrench to easily remove a nut from an FID collector without damaging the nut.
  • Unique, ergonomic handle that is easy to grip
  • Fits all instrument models
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty