Would you like to log in again?
Injection Port Repair Tool for Agilent Split/Splitless Injection Ports, Restek
Supplier: Avantor
21393
10852-872EA 156.77 USD InStock
10852-872
Injection Port Repair Tool for Agilent Split/Splitless Injection Ports, Restek
Tool Sets
The inlet seal at the base of a split/splitless injector allows a seal to form between the injection port and the inlet liner.
  • For use with Agilent split/splitless injection ports
This inlet seal wears over time and may become scratched or pitted, which compromises the sealing ability of the injector. Use the Restek injection port repair tool to easily resurface the inlet seal and remove contaminants; it saves time and money by preventing leaks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty