Would you like to log in again?
Male Pipe to Valco® Internal Adapter, Restek
Supplier: Restek
Valco®
20158 20159
10852-906EA 65.36 USD InStock
10852-906 10852-908
Male Pipe to Valco® Internal Adapter, Restek
Fittings and Connectors
These adapters make a minimum-volume connection from a female pipe fitting on a pressure gauge or regulator to a Valco® zero-dead-volume fitting.
  • Made of 300 series stainless steel; stainless steel ferrules included
Ideal for creating a zero-dead-volume fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty