Would you like to log in again?
Zero-Dead-Volume Valco® Internal Tee, Restek
Supplier: Restek
Valco®
20155 20154
10852-958EA 63.03 USD InStock
10852-958 10852-956
Zero-Dead-Volume Valco® Internal Tee, Restek
Fittings and Connectors
Use these tee connectors for making three-way connections with tubing.
  • 300 series stainless steel; stainless steel ferrules included
  • For 1/16" OD tubing
Designed so that no volume is introduced by the fitting which is not cleanly swept.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty