Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RESQ-Med™ Holster Set, Emergency Medical International

Supplier: Emergency Medical International
RESQ-Med™
688
10853-034KT 128.11 USD InStock
10853-034
RESQ-Med™ Holster Set, Emergency Medical International
First Aid Supplies First Aid Kits
RESQ-Med™ is the ultimate response holster set.

EMI has combined essential rescue equipment with critical medical supplies and equipment needed by the rescuer who is first on the scene. Extrication, Bleeding, CPR, and more. With RESQ-Med™ users are able to handle extreme conditions in the first few minutes of a rescue. The specially designed over-sized holster includes a front pocket for gloves, a water-repellant main compartment, and several instrument compartments. It is made of 1000 denier nylon with a quick release, Velcro-type belt loop. It fits belts up to 2.25" wide. The holster measures 21.6×12.7×6.9 cm (8.5×5×2.75").

RESQ-Med™ Holster set includes:

RESQ-Med™ Holster
4 Latex gloves
Thermal rescue blanket
CPR lifeshield
Instant ice pack, 10.2×12.7 cm (4×5")
Burn/blood stopper bandage
1 Sterile conforming bandage, 7.6 cm (3")
3 Sterile pads, 7.6×.6 cm (3×3")
Tourniquet
Life-Float II (Floating seatbelt cutter)
Window punch
Starlite waterproof flashlight
Deluxe EMS shears
Bandage scissor, 13.9 cm (5.5")

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty