Would you like to log in again?
ASI Varicella-Zoster, Arlington Scientific
Supplier: Arlington Scientific
840096AG
10853-068KT 431.89 USD InStock
10853-068
ASI Varicella-Zoster, Arlington Scientific
Assays ELISAs
ELISA Kit detects human antibodies to Varicella-Zoster Virus.
  • Qualitative and semiquantitative detection
  • Detects human IGG or IGM antibodies
ELISA for the qualitative and semiquantitative detection of human IGG/IGM antibodies to Varicella-Zoster virus.
Packaging: Kit of 96
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty