Would you like to log in again?
ASI Replacement HPLC Pump Piston Rods for Waters HPLC Systems, Restek
Supplier: Restek
25195 25196
10853-238EA 239.83 USD InStock
10853-238 10853-254
ASI Replacement HPLC Pump Piston Rods for Waters HPLC Systems, Restek
Chromatography Supplies Chromatography Pistons
Buffer salts and metals from the HPLC system become imbedded on the surface of the piston seal and gradually wear the surface of the piston rod.
Worn regions on the rod will quickly destroy the pump seals. In turn, the seal material gets trapped on the frit at the head of the column, which leads to increased system pressure and/or irregular or poor peak shape. As the piston continues to wear, a leak may develop at the pump. Any time piston seals are replaced, the piston rod should be inspected with a 10x magnifier or microscope to ensure it does not have wear or scratches. Replace the rod if you observe any glazed regions or scratches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty