Would you like to log in again?
MiniPulse Pulse Dampeners, Restek
Supplier: Restek
25238 25239
10803-378EA 728.58 USD InStock
10803-378 10853-354
MiniPulse Pulse Dampeners, Restek
Flow Meters
Improve system baseline stability while increasing the total system volume by only 160 µl.
  • Compact unit (2.5×1.5") can be placed almost anywhere
  • Small, 160 µl dead volume at atmospheric pressure
  • PEEK unit can withstand pressures to 5000 psi (34474 kPa)
  • 316 stainless steel unit can withstand pressures to 6000 psi (41369 kPa)
The MiniPulse pulse dampeners are ideal for applications where minimizing the total system volume is critical. Stainless steel and PEEK options for a wide range of applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty