Would you like to log in again?
Secure-Fit Fittings (Two Connectors), Restek
Supplier: RESTEK CORP
25211 26513
10853-390EA 173.66 USD InStock
10853-390 10853-604
Secure-Fit Fittings (Two Connectors), Restek
Fittings and Connectors
A good connection between HPLC components is necessary for trouble-free chromatography.
Secure-Fit connectors from Restek and Selerity Technologies ensure a consistent, leak-free seal—and they eliminate excess dead volume! An internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while you finger-tighten the nut.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty