Would you like to log in again?
FlatLine Pulse Damper, Restek
Supplier: RESTEK CORP
25340
10853-638EA 1034.73 USD InStock
10853-638
FlatLine Pulse Damper, Restek
Chromatography Supplies Chromatography Spare Parts
The ASI FlatLine Pulse Damper combines performance and reliability in a simple, easy-to-use housing.
  • Rupture-proof, no diaphragm—minimal risk of failure or leaks
  • Clean flush-out design—no sample carryover
  • Low internal volume—negligible effect on analyte bandwidth
Standard 10-32 inlet and outlet ports allow quick connection into virtually any HPLC system. Solid core technology provides reliable long-term operation without the downtime associated with ruptured or leaking membrane dampers. 600–700 µL internal volume at ambient pressure. Maximum pressure: 6,000 psi (41,369 kpa)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty