Would you like to log in again?
ValvTool Wrench, Restek
Supplier: Restek
25321
10853-760EA 73.24 USD InStock
10853-760
ValvTool Wrench, Restek
Wrenches
The ValvTool is a time-saving device that provides easy access to many hard-to-reach Rheodyne® or Valco® valves.
For 1/4" nuts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty