Would you like to log in again?
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Supplier: Restek
21787 21788
10854-094PK 34.05 USD InStock
10854-094 10854-096
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Vials
A variety of inserts compatible with screw-thread and crimp-top vials are available in a flat-bottomed format.
  • Screw-thread and crimp-top vial compatibility
  • Can also be used with snap seal vials
  • Polypropylene construction
Inserts for screw-thread, crimp-top, and snap seal vials are made of rugged polypropylene and feature a flat-bottomed design.
Ordering information: Polypropylene inserts available on request (1000-packs only). Also fit 4.0 ml Snap Seal vials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty