Would you like to log in again?
Big Mouth Step Screw-Thread Vials, Restek
Supplier: Restek
24628 24629 24627 24626
RK24629PK 345.7 USD InStock
RK24629 10854-346 10854-348 10854-344
Big Mouth Step Screw-Thread Vials, Restek
Vials
Big mouth step screw-thread vials feature a 2 ml volume and a 10 mm mouth.
  • 10 mm orifice
  • Screw-thread format
  • White graduated markings
  • Clear or amber color
These vials have a 2 ml capacity with obvious white marking spots.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty