Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Motic BA410 Elite Pathology Package
Supplier: Motic
1100100403047
10854-396EA 6455.96 USD InStock
10854-396
Motic BA410 Elite Pathology Package
Microscopes
The BA410 Elite Pathology Package is a complete pathology microscope bundle, which includes the Motic's flagship life-science microscope, the BA410 Elite.
  • Improved interpupillary distance and moveable eyepieces result in maximum comfort for any user
  • Rackless stage can hold two slides at once for quick comparison
  • Absence of gear rack on the stage allows for quick throughput of slides
  • High-end CCIS Achromatic optics minimize chromatic and spherical aberrations
  • A variety of optional upgrades and accessories is available
This package has been crafted to provide everything a pathologist or pathology educator needs to conduct their microscopy processes efficiently and effectively.
The BA410 Elite Pathology Package comes complete with all the accessories needed for pathology examination and training. A generous selection of add-ons, including a polarizer, gout.
Ordering information: Other accessories include a simple add on polarizer, Analyzer slider with rotating mount (360° rotatable), and Lambda/First Order Red Compensator. Microscope is customizable with various accessories. For all available options, please contact your local Avantor sales representative.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty