Would you like to log in again?
Male Quick Coupling, Parker, Restek
Supplier: Restek
21961 21958 21960 21957
10854-492EA 153.66 USD InStock
10854-492 10854-494 10855-534 10855-532
Male Quick Coupling, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Can be treated with Siltek®/Sulfinert or Silcosteel®-CR
Compatible with 1/4" or 1/8" openings depending on model.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty