Would you like to log in again?
Ball Valve, 2-Way, Straight, Parker, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
21589 21588
10854-526EA 591.2 USD InStock
10854-526 10854-528
Ball Valve, 2-Way, Straight, Parker, Restek
Valves Non Return Valves
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Parker manually, pneumatically, and electrically actuated two-way ball valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty