Would you like to log in again?
Straight Seamless 316L Grade Stainless Steel Tubing, Treated, Restek
Supplier: Restek
22901 22903 22899 22898 22902 22900
10854-858EA 437.46 USD InStock
10854-858 10854-902 10854-660 10854-662 10854-860 10854-664
Straight Seamless 316L Grade Stainless Steel Tubing, Treated, Restek
Tubing Transfer Tubing
Choose from 6-foot straight lengths of stainless steel tubing with outside diameters of 1/8" to 3/8".
  • Seamless stainless steel construction
  • Tubing can be cut to your exact requirements
  • Multiple lengths and different inside and outside diameters available
Lengths of stainless steel tubing are treated for optimal inertness and to extend lifetime.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty