Would you like to log in again?
Tube End Reducer, Swagelok®, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Swagelok®
22555 22589 22556 22591 23126 23128 23179 22590 22554 23176 23129 22553 23177 23178 23127 22588
10855-024EA 89.09 USD InStock
10855-024 10855-022 10058-954 10854-770 10854-650 10854-496 10854-652 10854-764 10854-766 10854-768 10854-636 10854-638 10854-828 10803-322 10803-320 10803-324
Tube End Reducer, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/16", 1/8", or 1/4" in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty