Would you like to log in again?
Ultra-High Purity Chrome-Plated Brass Line Gas Regulator, Restek
Supplier: Restek
22452 21666
10855-354EA 590.43 USD InStock
10855-354 RK21666
Ultra-High Purity Chrome-Plated Brass Line Gas Regulator, Restek
Gas Regulators
Use these ultra-high purity line gas regulators where you need to reduce the line pressure by 20 psig (138 kPa) or more.
  • Oxidation-resistant
  • Chrome-plated brass fabrication
  • Same purity protection as high-pressure cylinder regulators
Gas regulators feature 1/4" female NPT ports.
Ordering information: Order appropriate male connector, pipe-to-tube fittings.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty