Would you like to log in again?
Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with CGA Fittings, Restek
Supplier: Restek
20665 20666 20667
10855-370EA 1253.85 USD InStock
10855-370 10855-180 10855-622
Single-Stage Ultra-High Purity Stainless Steel Gas Regulators with CGA Fittings, Restek
Gas Regulators
UHP stainless steel regulators are the standard for ultra-high-purity and corrosion-resistant pressure regulation.
  • Identical gas purity protection as with dual-stage gas regulators
  • Use when there is secondary pressure regulation downstream
  • Also available with international fittings
All regulators are rated to 3000 psig maximum inlet pressure.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty