Would you like to log in again?
Elbow, Swagelok®, Restek
Supplier: Restek
Swagelok®
22549 22585 22550 22584
10855-496EA 126.78 USD InStock
10855-496 10855-494 10855-508 10855-506
Elbow, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Elbow fittings are hot forged and machined. Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty