Would you like to log in again?
Cross, Swagelok®, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Swagelok®
23125 23174 22551 23124 22587 22586 23175 22552
10058-908PK 90.19 USD InStock
10058-908 10855-498 10058-952 10855-656 10855-512 10058-906 10855-500 10855-510
Cross, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Use these cross connectors for making four-way connections with tubing. Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty