Would you like to log in again?
Swagelok® VCO O-Ring Face Seal Fittings, Treated, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Swagelok®
23209 23207 23215 23214
10854-754PK 63.18 USD InStock
10854-754 10854-756 10855-584 10855-524
Swagelok® VCO O-Ring Face Seal Fittings, Treated, Restek
Fittings and Connectors
Swagelok VCO O-ring face seal fittings are designed for rapid assembly in pipe, tube, and welded systems.
  • Unique design allows easy installation where space is limited
  • Assemblies can be used across a wide range of temperatures
  • Smooth finish on gland face ensures positive seal
  • Sealing is accomplished with a captive O-ring in the body component
Assemblies can be used from high pressure to critical vacuum across a wide range of temperatures.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty