Would you like to log in again?
Male Connector, Swagelok®, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Swagelok®
23186 23133 22561 22565 23136 22595 22563 22611 22596 23185 22564 22912 23184 23135 23134 22610 22612 22907 22562 23183 23187 23137
10855-530EA 70.28 USD InStock
10855-530 10854-480 10855-052 10058-910 10854-482 10854-484 10854-880 10854-472 10854-882 10854-772 10854-486 10854-774 10854-884 10854-776 10854-632 10854-634 10854-876 10854-912 10058-912 10058-956 10854-906 10854-908
Male Connector, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available in 1/16" to 1/4" NPT sizing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty