Would you like to log in again?
Dual-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Supplier: Restek
22140 22141 22377 22378 22376 22379
10855-540EA 1469.96 USD InStock
10855-540 10855-582 10855-580 10855-470 10855-538 10855-536
Dual-Stage Ultra-High Purity Stainless Steel Gas Regulators with International Fittings, Restek
Gas Regulators
UHP stainless steel regulators are the standard for ultra-high-purity and corrosion-resistant pressure regulation.
  • Secondary pressure regulation not needed
  • Most stable outlet pressure control
  • Also available with CGA fittings
All regulators are rated to 3000 psig maximum inlet pressure.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty