Would you like to log in again?
Plug, Swagelok®, Restek
Supplier: Restek
Swagelok®
23113 22573 22574 22597 23163 22620 23114 23112 23162 23164
10855-564EA 61.96 USD InStock
10855-564 10855-562 10855-560 10855-708 10058-930 10058-932 10058-888 10855-638 10855-558 10855-710
Plug, Swagelok®, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available in sizes of 1/16", 1/8", or 1/4" in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty