Would you like to log in again?
Cross, Parker, Restek
Supplier: Restek
22872 21930 21829 21543 22873 21542 21929 21830
10855-586EA 161.58 USD InStock
10855-586 10855-010 RK21829 10855-734 10855-798 10855-732 10855-588 10855-796
Cross, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Cross fitting allows for four connections with 1/8" openings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty