Would you like to log in again?
Port Connector, Parker, Restek
Supplier: Restek
22886 21548 21935 22885 21936 22884 21550 21937 21836 21549
10855-652EA 72.51 USD InStock
10855-652 10855-650 10855-184 10855-590 10855-606 10854-490 10855-604 10855-428 10855-248 10855-246
Port Connector, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty