Would you like to log in again?
Ball Valve, Parker Balston® Shut-Off Gas Valve, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22192 22195 22193 22194
10855-718EA 234.15 USD InStock
10855-718 10855-636 10855-720 10855-016
Ball Valve, Parker Balston® Shut-Off Gas Valve, Restek
Valves Non Return Valves
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
Ball valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications. Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty