Would you like to log in again?
Toggle Valve, Swagelok® Shut-Off Gas Valve, Restek
Supplier: Restek
23198 23142 23199 23143
10803-330EA 219.01 USD InStock
10803-330 10855-728 10855-702 10855-730
Toggle Valve, Swagelok® Shut-Off Gas Valve, Restek
Valves Air Regulation Valves
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
Toggle valves provide 1/4 turn for on-off control of fluids utilized in process and instrumentation applications. Available with connection sizes of 1/8" or 1/4" in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty