Would you like to log in again?
Tee, Parker, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
21927 21928 22868 21828 21926 21827 21826 22866 22527 22526 22528 22867
10855-772EA 104.83 USD InStock
10855-772 10855-770 10855-790 10855-826 RK21827 10855-814 10855-648 10855-824 10855-812 10855-700 10855-810 10855-752
Tee, Parker, Restek
Fittings and Connectors
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR
Use these tee connectors for making three-way connections with tubing. Available with connection sizes of 1/16", 1/8", or 1/4" in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty