Would you like to log in again?
Swagelok® Fitting Kit, Stainless Steel, Restek
Supplier: Restek
Swagelok®
23197
10855-830KT 1879.91 USD InStock
10855-830
Swagelok® Fitting Kit, Stainless Steel, Restek
Fittings and Connectors
The Swagelok® fitting kit includes the most common assortment of stainless steel fittings for GC fixtures.
  • Parts list makes reordering easy
  • Parts come in a sturdy toolbox for easy and convenient storage
Common assortment of fittings are delivered in a rugged storage system with compartments for easy identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty