Would you like to log in again?
Cylinder Valve Wrench, Restek
Supplier: Restek
21321
10855-832EA 147.95 USD InStock
10855-832
Cylinder Valve Wrench, Restek
Wrenches
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel.
It is also suitable for removing difficult cylinder caps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty