Would you like to log in again?
Merlin Microseal™ Septa for PerkinElmer GCs, Restek
Supplier: Restek
22781
10855-838EA 497.62 USD InStock
10855-838
Merlin Microseal™ Septa for PerkinElmer GCs, Restek
Septa
The advantages of the Merlin Microseal septum include elimination of septum coring, longer life, and consistent low needle insertion force.
  • 400 °C max injection port temperature
  • Longer lifetime
  • Installation is simple
The Microseal septum incorporates two separate sealing mechanisms. These sliding seals eliminate septum coring and the resulting accumulation of septum crumbs in the injection port liner. The Microseal septum uses a 23-gauge (0.63 mm, 0.025") needle or probe with a blunt, truncated conical tip. Since the syringe plunger end details are determined by manual or autosampler compatibility, often a removable needle syringe is an effective way to match both of these requirements. Installation is simple, requiring no modification of the injection port.
Delivery information: Includes Nut, Adapter, O-Ring and 2 General-Purpose (#410) Microseals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty