Would you like to log in again?
EZ No-Vent® GC Column-Mass Spectrometer Connector, Restek
Supplier: Restek
EZ No-Vent®
22431
10856-020EA 272.35 USD InStock
10856-020
EZ No-Vent® GC Column-Mass Spectrometer Connector, Restek
Fittings and Connectors
Restek has designed the EZ No-Vent® GC column-mass spectrometer connector to be simple and easy to use.
  • Change GC-MS columns in minutes without venting
  • Easy to install and maintain
  • Gold-plated body for inertness
  • High-temperature polyimide ferrules eliminate leaks at the problematic transfer line fitting
  • Lower cost than other "no-vent" fittings
A critical orifice in the EZ No-Vent® connector minimizes the amount of oxygen allowed into the MS source, eliminating the need for purge gas as is required for other manufacturer's vent systems. The lengthy vent and pump-down cycle is not required, and normal instrument down-time from a column change is dramatically reduced. The EZ No-Vent® connector easily attaches to the MS source without special tools or extra plumbing.
Accessories information: For EZ No-Vent® GC column-mass spectrometer connectors and 100 μm deactivated transfer line.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty