Would you like to log in again?
EZ No-Vent® GC Column-Mass Spectrometer Connector for Mass Spectrometers, Restek
Supplier: Restek
EZ No-Vent®
22454
10856-024EA 325.11 USD InStock
10856-024
EZ No-Vent® GC Column-Mass Spectrometer Connector for Mass Spectrometers, Restek
Fittings and Connectors
The EZ No-Vent® GC column-mass spectrometer connector is designed to be simple and easy to use.
  • Change GC-MS columns in minutes without venting
  • Easy to install and maintain
  • Gold-plated body for inertness
  • High-temperature polyimide ferrules eliminate leaks at the problematic transfer line fitting
  • Lower cost than other "no-vent" fittings
A critical orifice in the EZ No-Vent® connector minimizes the amount of oxygen allowed into the MS source, eliminating the need for purge gas as is required for other manufacturers' vent systems. This enables you to skip the lengthy vent and pump-down cycle otherwise required when you make a column change, saving nearly a day of downtime with each column change. The EZ No-Vent® connector easily attaches to the MS source without special tools or extra plumbing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty