Would you like to log in again?
Extended Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Supplier: Restek
22634 22637 22635 22636 22633
10856-044EA 65.15 USD InStock
10856-044 10856-040 10856-042 10856-036 10856-038
Extended Capillary Column Nuts for Agilent 5890/6850/6890/7890 GCs, Restek
Fittings and Connectors
Visible below inlet insulating cup for easy, breakage-free column installation without removing cup.
  • Finger-tight design to allow wrench-free use
  • Available for standard (1/16") and compact (Agilent-style) ferrules
  • Made from high-quality stainless steel
  • Meets or exceeds original manufacturer's performance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty