Would you like to log in again?
Inlet Adapter Kit for Dual Column Installation for Agilent Capillary Injectors, Split/Splitless Fitting for Capillary Columns, Restek
Supplier: Restek
27185
10856-162EA 108.74 USD InStock
10856-162
Inlet Adapter Kit for Dual Column Installation for Agilent Capillary Injectors, Split/Splitless Fitting for Capillary Columns, Restek
Fittings and Connectors
Capillary columns are easier to install due to the nut protruding farther from the insulated injection port chamber.
  • 1/16-inch split/splitless fitting that accepts standard, two-hole capillary ferrules
  • Same column insertion depth as the original manufacturer's equipment
For use with Agilent split/splitless fittings for capillary columns.
Kit includes adapter fitting, 1/16" capillary nut, gold-plated 1.2 mm ID dual Vespel ring inlet seal, one 0.4 mm ID two-hole ferrule, and one 0.5 mm ID two-hole ferrule.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty