Would you like to log in again?
Dualmat™ Static Dissipative Mats, Bulk Roll, Dark Gray, ACL
Supplier: ACL Staticide
8385DGYR3640 8385DGYR2440 8385DGYR3040 8385DGYR4840
10856-352EA 1 USD InStock
10856-352 10856-350 10856-348 10856-346
Dualmat™ Static Dissipative Mats, Bulk Roll, Dark Gray, ACL
Surface Protectors Bench Protectors
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer, bulk roll mats are supplied in 40 foot lengths. They are available in 24", 30", 36", and 48" widths. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes it ideal for use with constant monitors. Embossed finish reduces glare, Resistance is 10⁶ to 10⁸ ohms. Electricals warranted for the lifetime of the mat.
Certifications: Compliant to ANSI/ESD S20.20 and RoHS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty