Would you like to log in again?
BT Lab Systems Micro-Plate Shaker, G-Biosciences
Supplier: G-Biosciences
BT908
10856-394EA 1441.76 USD InStock
10856-394
BT Lab Systems Micro-Plate Shaker, G-Biosciences
Shakers and Mixers Microplate Shakers
Micro-Plate Shaker, Ideal for ELISA plates (96 and 384 wells) and cell culture plates
  • Speed and time under microprocessor control
  • Safe and stable with minimal noise
  • DC brushless motor, long life, free of maintenance. Can provide gentle and powerful mixing
  • Timing function, alarm notification after program completion
Micro-Plate Shaker has LED digital display with simple and easy operation panel that can accurately display the time and the speed of shaking parameters.
Can accommodate 4 standard micro plates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty