Would you like to log in again?
Polycarbonate Face Shields, Oberon
Supplier: Oberon
171R 179R
10856-540PK 380.2 USD InStock
10856-540 87001-674
Polycarbonate Face Shields, Oberon
Face Shields
The clear UV Protective Series face shields provide 100% protection from UV energy from 200 to 400nm.
  • Scratch resitant outside
  • Antifog inside
Not suitable for electrical arc flash protection, not intended for high intensity laser type applications.
Certifications: Meets ANSI Z87.1-2010 standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty